Anisomycin ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2R,3S,4S)-4-Hydroxy-2-(4-methoxybenzyl)pyrrolidin-3-yl acetate
A646979-10MG
A646979-1MG
A646979-50MG
77627-290EA
42.23
USD
77627-290
77627-292
77627-294
Anisomycin ≥98%
Anisomycin
Caution: for Research Use Only
Formula:
C₁₄H₁₉NO₄ MW: 265.31 g/mol |
MDL Number:
MFCD00077650 CAS Number: 22862-76-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...