(5-Isopropoxypyridin-3-yl)boronic acid 95+%

Supplier: Ambeed
Warning

A633723-100MG A633723-1G A633723-250MG
77884-494EA 47 USD
77884-494 77884-495 77884-496
(5-Isopropoxypyridin-3-yl)boronic acid 95+%
(5-Isopropoxypyridin-3-yl)boronic acid
Formula: C₈H₁₂BNO₃
MDL Number: MFCD15529675
CAS Number: 850991-41-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR