(3S,6S)-5-(tert-Butoxycarbonyl)-1,1-difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A606553-1G
77572-970EA 942 USD
77572-970
(3S,6S)-5-(tert-Butoxycarbonyl)-1,1-difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid ≥97%
(3S,6S)-5-(tert-Butoxycarbonyl)-1,1-difluoro-5-azaspiro[2.4]heptane-6-carboxylic acid

Caution: for Research Use Only

Formula: C₁₂H₁₇F₂NO₄
MW: 277.2645 g/mol
MDL Number: MFCD28396445
CAS Number: 1980007-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR