(5-Acetylpyridin-3-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A555008-1G A555008-250MG A555008-100MG A555008-5G
77686-427EA 131 USD
77686-427 77686-428 77901-104 77901-505
(5-Acetylpyridin-3-yl)boronic acid ≥95%
(5-Acetylpyridin-3-yl)boronic acid

Caution: for Research Use Only

Formula: C₇H₈BNO₃
MDL Number: MFCD10696658
CAS Number: 1033745-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR