n-Octacosane ≥98%

Supplier: Ambeed
Danger

Synonyms: N-octacosane

A553283-5G A553283-100G A553283-1G A553283-25G
77602-342EA 28.7 USD
77602-342 77669-438 77669-440 77880-192
n-Octacosane ≥98%
n-Octacosane

Caution: for Research Use Only

Formula: H₃C(CH₂)₂₆CH₃
MW: 394.77 g/mol
Boiling Pt: 432
MDL Number: MFCD00009355
CAS Number: 630-02-4
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR