Flavokawain B ≥98%

Supplier: Ambeed
Warning

Synonyms: (E)-1-(2-Hydroxy-4,6-dimethoxyphenyl)-3-phenylprop-2-en-1-one, Flavokavain B

A522040-5MG A522040-100MG A522040-10MG A522040-50MG
77669-310EA 89 USD
77669-310 77872-876 77872-877 77872-878
Flavokawain B ≥98%
Flavokawain B

Caution: for Research Use Only

Formula: C₁₇H₁₆O₄
MW: 284.32 g/mol
MDL Number: MFCD00075877
CAS Number: 1775-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR