(6-Methoxybenzofuran-2-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

A500503-100MG A500503-250MG A500503-5G
77891-013EA 58 USD
77891-013 77891-014 77891-015
(6-Methoxybenzofuran-2-yl)boronic acid ≥97%
(6-Methoxybenzofuran-2-yl)boronic acid

Caution: for Research Use Only

Formula: C₉H₉BO₄
MDL Number: MFCD18250058
CAS Number: 952737-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR