(S)-(+)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥98%

Supplier: Ambeed
Warning

A485957-250MG A485957-100MG A485957-1G
77572-538EA 94 USD
77572-538 77617-106 77747-000
(S)-(+)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl ≥98%
(S)-(+)-2,2'-Bis[di(3,5-di-i-propyl-4-dimethylaminophenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

Caution: for Research Use Only

Formula: C₇₀H₁₀₀N₄O₂P₂
MDL Number: MFCD09753010
CAS Number: 919338-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR