5-Chlorovaleronitrile ≥98%

Supplier: Ambeed
Danger

Synonyms: 5-Chloropentanenitrile

A473973-100G A473973-25G A473973-5G
77598-506EA 48.18 USD
77598-506 77598-508 77626-944
5-Chlorovaleronitrile ≥98%
5-Chlorovaleronitrile

Caution: for Research Use Only

Formula: C₅H₈ClN
MDL Number: MFCD00001977
CAS Number: 6280-87-1
UN: 3276
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR