Cefotaxime sodium salt ≥98%

Supplier: Ambeed
Danger

Synonyms: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

A468233-1MG A468233-250MG A468233-25G A468233-1G A468233-5G
77673-196EA 5 USD
77673-196 77884-199 77884-200 77884-198 77884-201
Cefotaxime sodium salt ≥98%
Cefotaxime sodium salt

Caution: for Research Use Only

Formula: C₁₆H₁₇N₅O₇S₂
MW: 477.44 g/mol
MDL Number: MFCD00079073
CAS Number: 64485-93-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR