(2-Hydroxy-4-methylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A452742-100MG A452742-1G A452742-250MG A452742-5G
77890-919EA 132 USD
77890-919 77890-920 77890-921 77890-922
(2-Hydroxy-4-methylphenyl)boronic acid ≥98%
(2-Hydroxy-4-methylphenyl)boronic acid

Caution: for Research Use Only

Formula: C₇H₉BO₃
MDL Number: MFCD18397304
CAS Number: 259209-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR