Hygromycin B ≥950 U/mg

Supplier: Ambeed
Danger

Synonyms: (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol

A446642-250MG A446642-1MG A446642-100MG A446642-1G
77601-938EA 62.53 USD
77601-938 77669-004 77879-976 77879-977
Hygromycin B ≥950 U/mg
Hygromycin B

Caution: for Research Use Only

Formula: C₂₀H₃₇N₃O₁₃
MW: 527.5 g/mol
MDL Number: MFCD06795479
CAS Number: 31282-04-9
UN: 3462
ADR: 6.1 Ⅰ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR