7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Dioxopyrrolidin-1-yl 7-(diethylamino)-2-oxo-2H-chromene-3-carboxylate, DEAC, DNP-X acid, SE
A437996-100MG
A437996-1G
A437996-250MG
77601-902EA
141.45
USD
77601-902
77626-874
77878-193
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester 98+%
7-(Diethylamino)coumarin-3-carboxylic acid, N-succinimidyl ester
Caution: for Research Use Only
Formula:
C₁₈H₁₈N₂O₆ MW: 358.35 g/mol |
MDL Number:
MFCD00467585 CAS Number: 139346-57-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...