Zinc diethyldithiocarbamate ≥99%

Supplier: Ambeed
Danger

Synonyms: Zinc dethyldithiocarbamate, Diethyldithiocarbamic Acid Zinc Salt

A442461-25G A442461-100G
77594-384EA 5.13 USD
77594-384 77601-918
Zinc diethyldithiocarbamate ≥99%
Zinc diethyldithiocarbamate

Caution: for Research Use Only

Formula: C₅H₁₁NS₂
MW: 361.90 g/mol
MDL Number: MFCD00064798
CAS Number: 14324-55-1
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR