(4-Fluoropyridin-2-yl)boronic acid 95+%
Supplier: Ambeed
|
Warning
|
A406664-1G
A406664-100MG
77572-258EA
260
USD
77572-258
77886-086
(4-Fluoropyridin-2-yl)boronic acid 95+%
(4-Fluoropyridin-2-yl)boronic acid
Caution: for Research Use Only
Formula:
C₅H₅BFNO₂ |
MDL Number:
MFCD08061577 CAS Number: 1208101-73-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...