Josamycin ≥920 IU/mg

Supplier: Ambeed
Warning

Synonyms: (2S,3S,4R,6S)-6-(((2R,3S,4R,5R,6S)-6-(((4R,5S,6S,7R,9R,10R,11E,13E,16R)-4-Acetoxy-10-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-7-(2-oxoethyl)oxacyclohexadeca-11,13-dien-6-yl)oxy)-4-(dimethylamino)-5-hydroxy-2-methyltetrahydro-2H-pyran-3-yl)oxy)-4-hydroxy-2,4-

A387439-1G A387439-250MG
77879-833EA 338 USD
77879-833 77879-834
Josamycin ≥920 IU/mg
Josamycin

Caution: for Research Use Only

Formula: C₄₂H₆₉NO₁₅
MDL Number: MFCD00211043
CAS Number: 16846-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR