Flt-3 inhibitor 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-dimethoxybenzamido)-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxamide, TCS 359, TCS-359

A374354-10MG A374354-1MG A374354-5MG
77613-122EA 36.05 USD
77613-122 77613-124 77613-126
Flt-3 inhibitor 98+%
Flt-3 inhibitor

Caution: for Research Use Only

Formula: C₁₈H₂₀N₂O₄S
MW: 360.43 g/mol
MDL Number: MFCD00617269
CAS Number: 301305-73-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR