(2-methylnaphthalen-1-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A316945-1G A316945-100MG A316945-250MG A316945-5G
77593-882EA 61.5 USD
77593-882 77890-628 77890-629 77890-630
(2-methylnaphthalen-1-yl)boronic acid ≥98%
(2-methylnaphthalen-1-yl)boronic acid

Caution: for Research Use Only

Formula: C₁₁H₁₁BO₂
MDL Number: MFCD03452758
CAS Number: 103989-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR