Ethyl methoxyacetate ≥98%

Supplier: Ambeed
Danger

Synonyms: Methoxyacetic ethyl ester

A307561-25G A307561-100G A307561-5G
77593-844EA 16.4 USD
77593-844 77668-360 77668-362
Ethyl methoxyacetate ≥98%
Ethyl methoxyacetate

Caution: for Research Use Only

Formula: C₅H₁₀O₃
MW: 118.13 g/mol
Boiling Pt: 44
MDL Number: MFCD00009176
CAS Number: 3938-96-3
UN: 3272
ADR: 3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR