N2,9-Diacetylguanine ≥96%

Supplier: Ambeed
Warning

Synonyms: N,9-Diacetylguanine, 2-Acetamido-9-acetyl-6-oxopurine, N-(9-Acetyl-6-oxo-6,9-dihydro-1H-purin-2-yl)acetamide

A289916-100G A289916-25G
77626-498EA 30.9 USD
77626-498 77626-500
N2,9-Diacetylguanine ≥96%
N2,9-Diacetylguanine

Caution: for Research Use Only

Formula: C₉H₉N₅O₃
MW: 235.20 g/mol
MDL Number: MFCD00142116
CAS Number: 3056-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR