Astragaloside A ≥98%

Supplier: Ambeed
Warning

Synonyms: Astragaloside IV, (2R,3R,4S,5S,6R)-2-(((2aR,3R,4S,5aS,5bS,7S,7aR,9S,11aR,12aS)-4-Hydroxy-3-((2S,5R)-5-(2-hydroxypropan-2-yl)-2-methyltetrahydrofuran-2-yl)-2a,5a,8,8-tetramethyl-9-(((2S,3R,4S,5R)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl)oxy)hexadecahydrocyclopenta[a]cyclopropa[e]phenanthren-7-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A287152-10MG A287152-50MG A287152-5MG
77593-758EA 7.18 USD
77593-758 77593-760 77593-762
Astragaloside A ≥98%
Astragaloside A

Caution: for Research Use Only

Formula: C₄₁H₆₈O₁₄
MW: 784.970 g/mol
MDL Number: MFCD00210476
CAS Number: 83207-58-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR