(6-(Isopropylthio)pyridin-3-yl)boronic acid ≥98%

Supplier: Ambeed
Warning

A255837-1G A255837-250MG
77885-667EA 101 USD
77885-667 77885-668
(6-(Isopropylthio)pyridin-3-yl)boronic acid ≥98%
(6-(Isopropylthio)pyridin-3-yl)boronic acid

Caution: for Research Use Only

Formula: C₈H₁₂BNO₂S
MDL Number: MFCD16295060
CAS Number: 1256345-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR