(2-Bromo-6-fluoro-3-isopropoxyphenyl)boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A245280-1G
A245280-250MG
A245280-5G
77672-194EA
15
USD
77672-194
77883-655
77883-656
(2-Bromo-6-fluoro-3-isopropoxyphenyl)boronic acid ≥98%
(2-Bromo-6-fluoro-3-isopropoxyphenyl)boronic acid
Caution: for Research Use Only
Formula:
C₉H₁₁BBrFO₃ |
MDL Number:
MFCD08276826 CAS Number: 1072951-75-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...