2-Chloro-1H-indole-3-carbaldehyde ≥97%

Supplier: Ambeed
Warning

A240444-1G A240444-5G
77892-423EA 49 USD
77892-423 77892-424
2-Chloro-1H-indole-3-carbaldehyde ≥97%
2-Chloro-1H-indole-3-carbaldehyde

Caution: for Research Use Only

Formula: C₉H₆ClNO
MW: 179.61 g/mol
MDL Number: MFCD00111620
CAS Number: 5059-30-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR