2-Chloro-6-methylpyridine-4-boronic acid ≥98%

Supplier: Ambeed
Warning

A223329-1G A223329-250MG A223329-5G
77890-318EA 219 USD
77890-318 77890-319 77890-320
2-Chloro-6-methylpyridine-4-boronic acid ≥98%
2-Chloro-6-methylpyridine-4-boronic acid

Caution: for Research Use Only

Formula: C₆H₇BClNO₂
MDL Number: MFCD13188704
CAS Number: 1320397-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR