(5-Amino-2-methylphenyl)boronic acid ≥98%

Supplier: Ambeed
Warning

A206451-250MG A206451-100MG A206451-1G
77685-532EA 63 USD
77685-532 77900-840 77902-017
(5-Amino-2-methylphenyl)boronic acid ≥98%
(5-Amino-2-methylphenyl)boronic acid

Caution: for Research Use Only

Formula: C₇H₁₀BNO₂
MDL Number: MFCD22192389
CAS Number: 1003042-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR