1-Hexacosanol ≥90%

Supplier: Ambeed
Warning

Synonyms: 1-Hydroxy hexacosane, 1-Hexacosyl alcohol, Cerotyl alcohol, Pentacosyl carbinol

A1978535-1G A1978535-250MG A1978535-5G A1978535-100MG A1978535-25G
77598-284EA 10.25 USD
77598-284 77598-286 77598-288 77598-282 77877-576
1-Hexacosanol ≥90%
1-Hexacosanol

Caution: for Research Use Only

Formula: H₃C(CH₂)₂₄CH₂OH
MW: 382.71 g/mol
Boiling Pt: 412
MDL Number: MFCD00002940
CAS Number: 506-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR