N-Methylanthranilic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: 2-(Methylamino)benzoic acid

A188996-100G A188996-5G
77667-572EA 171 USD
77667-572 77879-213
N-Methylanthranilic acid ≥99%
N-Methylanthranilic acid

Caution: for Research Use Only

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 80
Melting Pt: 172
MDL Number: MFCD00002424
CAS Number: 119-68-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR