bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥98%

Supplier: Ambeed
Warning

A186328-100MG A186328-250MG A186328-50MG
77879-203EA 64 USD
77879-203 77879-204 77879-205
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) ≥98%
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

Caution: for Research Use Only

Formula: C₂₇H₂₈N₆O·3HCl
MDL Number: MFCD00012679
CAS Number: 23491-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR