Choline dihydrogen citrate ≥97%

Supplier: Ambeed
Warning

A179663-100G A179663-500G
77625-850EA 9.27 USD
77625-850 77879-178
Choline dihydrogen citrate ≥97%
Choline dihydrogen citrate

Caution: for Research Use Only

Formula: C₁₁H₂₁NO₈
MDL Number: MFCD00038734
CAS Number: 77-91-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR