Polyvinyl alcohol 2099

Supplier: Ambeed
Warning

A1495841-1000G A1495841-500G A1495841-100G
77597-720EA 13.33 USD
77597-720 77597-722 77624-572
Polyvinyl alcohol 2099
Polyvinyl alcohol

Caution: for Research Use Only

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR