Polyvinyl alcohol 1799

Supplier: Ambeed
Warning

A1495839-1000G A1495839-100G A1495839-500G
77597-714EA 13.33 USD
77597-714 77597-716 77597-718
Polyvinyl alcohol 1799
Polyvinyl alcohol

Caution: for Research Use Only

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR