Polyvinyl alcohol 1788

Supplier: Ambeed
Warning

A1495835-1000G A1495835-100G A1495835-500G
77597-704EA 12.3 USD
77597-704 77597-706 77597-708
Polyvinyl alcohol 1788
Polyvinyl alcohol

Caution: for Research Use Only

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR