Polyvinyl alcohol 2488

Supplier: Ambeed
Warning

A1495833-100G A1495833-500G
77597-702EA 8.2 USD
77597-702 77875-636
Polyvinyl alcohol 2488
Polyvinyl alcohol

Caution: for Research Use Only

Formula: [-CH₂CH(OH)-]n
MDL Number: MFCD00081922
CAS Number: 9002-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR