Mupirocin 920-1020 μg/mg

Supplier: Ambeed
Warning

Synonyms: 9-(((E)-4-((2S,3R,4R,5S)-3,4-Dihydroxy-5-(((2S,3S)-3-((2S,3S)-3-hydroxybutan-2-yl)oxiran-2-yl)methyl)tetrahydro-2H-pyran-2-yl)-3-methylbut-2-enoyl)oxy)nonanoic acid

A1477160-1G A1477160-25G A1477160-5G
77622-238EA 17.51 USD
77622-238 77622-240 77622-242
Mupirocin 920-1020 μg/mg
Mupirocin

Caution: for Research Use Only

Formula: C₂₆H₄₄O₉
MW: 500.629 g/mol
MDL Number: MFCD01711620
CAS Number: 12650-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR