Hydroxyethyl cellulose, 1500~2500 mPa.s, 25℃

Supplier: Ambeed
Warning

A1460276-500G A1460276-1000G A1460276-100G A1460276-25G
77624-220EA 13.39 USD
77624-220 77874-959 77874-960 77874-961
Hydroxyethyl cellulose, 1500~2500 mPa.s, 25℃
Hydroxyethyl cellulose

Caution: for Research Use Only

MDL Number: MFCD00072770
CAS Number: 9004-62-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR