Sudan I ≥98% (by HPLC)
Supplier: Ambeed
|
Warning
|
Synonyms:
Scarlet B, 1-(Phenyldiazenyl)naphthalen-2-ol, 1-Phenylazo-2-naphthol, Oil Orange, Solvent Yellow 14, Sudan Yellow
A1459875-25G
A1459875-100G
77597-286EA
8.2
USD
77597-286
77624-218
Sudan I ≥98% (by HPLC)
Sudan I
Caution: for Research Use Only
Formula:
C₁₆H₁₂N₂O MW: 248.28 g/mol |
MDL Number:
MFCD00003911 CAS Number: 842-07-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...