Solvent red 197, Stain grade

Supplier: Ambeed
Warning

Synonyms: 3-(Diethylamino)-7-imino-7H-benzo[4,5]imidazo[1,2-a]chromeno[3,2-c]pyridine-6-carbonitrile

A1457369-100G A1457369-25G A1457369-500G
77597-276EA 41 USD
77597-276 77624-210 77624-212
Solvent red 197, Stain grade
Solvent red 197

Caution: for Research Use Only

Formula: C₂₃H₁₉N₅O
MW: 381.44 g/mol
MDL Number: MFCD02176212
CAS Number: 52372-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR