Ethyl oleate 75+% (by GC)

Supplier: Ambeed
Warning

Synonyms: Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester

A1453636-1000G A1453636-25G A1453636-100G A1453636-500G
77622-162EA 41.2 USD
77622-162 77622-164 77874-924 77874-925
Ethyl oleate 75+% (by GC)
Ethyl oleate

Caution: for Research Use Only

Formula: C₂₀H₃₈O₂
MDL Number: MFCD00009579
CAS Number: 111-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR