N,2,6-Trimethylaniline ≥95%

Supplier: Ambeed
Warning

A144632-1G A144632-250MG
77881-124EA 348 USD
77881-124 77881-125
N,2,6-Trimethylaniline ≥95%
N,2,6-Trimethylaniline

Caution: for Research Use Only

Formula: C₉H₁₃N
MDL Number: MFCD00613418
CAS Number: 767-71-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR