Diosgenin ≥98%

Supplier: Ambeed
Warning

Synonyms: Nitogenin, (2'R,4S,5'R,6aR,6bS,8aS,8bR,9S,11aS,12aS,12bS)-5',6a,8a,9-Tetramethyl-1,3,3',4,4',5,5',6,6a,6b,6',7,8,8a,8b,9,11a,12,12a,12b-icosahydrospiro[naphtho[2',1':4,5]indeno[2,1-b]furan-10,2'-pyran]-4-ol

A136414-100G A136414-25G A136414-500G A136414-5G A136414-10MG A136414-50MG
77592-578EA 75.85 USD
77592-578 77592-580 77592-582 77592-584 77670-954 77670-956
Diosgenin ≥98%
Diosgenin

Caution: for Research Use Only

Formula: C₂₇H₄₂O₃
MW: 414.63 g/mol
MDL Number: MFCD00016887
CAS Number: 512-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR