(3-((Methylamino)methyl)phenyl)boronic acid ≥95%

Supplier: Ambeed

A132679-100MG A132679-250MG
77889-902EA 53 USD
77889-902 77889-903
(3-((Methylamino)methyl)phenyl)boronic acid ≥95%
(3-((Methylamino)methyl)phenyl)boronic acid

Caution: for Research Use Only

Formula: C₈H₁₂BNO₂
MDL Number: MFCD18453453
CAS Number: 1146614-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR