GDC-046 ≥98%

Supplier: Ambeed
Warning

Synonyms: 2,6-Dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide

A1325556-100MG A1325556-250MG
77867-278EA 231 USD
77867-278 77867-279
GDC-046 ≥98%
GDC-046

Caution: for Research Use Only

Formula: C₁₆H₁₃Cl₂N₃O₂
MDL Number: MFCD28155087
CAS Number: 1258292-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR