Cefoxitin sodium ≥98%

Supplier: Ambeed
Warning

Synonyms: Mefoxin, Cefoxitin sodium salt, Sodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

A125671-1G A125671-100G A125671-100MG A125671-250MG A125671-25G
77599-816EA 17.43 USD
77599-816 77624-048 77624-050 77624-052 77878-922
Cefoxitin sodium ≥98%
Cefoxitin sodium

Caution: for Research Use Only

Formula: C₁₆H₁₆N₃NaO₇S₂
MW: 449.428 g/mol
MDL Number: MFCD00079042
CAS Number: 33564-30-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR