Wortmannin ≥98%

Supplier: Ambeed
Warning

Synonyms: (1S,6bR,9aS,11R,11bR)-1-(Methoxymethyl)-9a,11b-dimethyl-3,6,9-trioxo-1,6,6b,7,8,9,9a,10,11,11b-decahydro-3H-furo[4,3,2-de]indeno[4,5-h]isochromen-11-yl acetate

A117244-10MG A117244-50MG A117244-100MG A117244-250MG A117244-5MG
77666-686EA 109.38 USD
77666-686 77666-688 77871-494 77871-495 77871-496
Wortmannin ≥98%
Wortmannin

Caution: for Research Use Only

Formula: C₂₃H₂₄O₈
MDL Number: MFCD00133927
CAS Number: 19545-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR