Fusidate sodium ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A110028-100MG A110028-250MG A110028-25G A110028-5G A110028-1G
77683-008EA 15 USD
77683-008 77683-009 77683-010 77683-011 77885-008
Fusidate sodium ≥98%, Ambeed.Inc

Caution: For research use only.

Formula: C₃₁H₄₇NaO₆
MDL Number: MFCD09054714
CAS Number: 751-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR