Human Recombinant PRKG2 (GST tag), Active (from Baculovirus-Insect Cells)
Supplier: Sino Biological
P79-10G-10 P79-10G-50 P79-10G-500
77562-320EA 348 USD
77562-320 77562-322 77562-324
Human Recombinant PRKG2 (GST tag), Active (from Baculovirus-Insect Cells)
Proteins and Peptides
Recombinant full length human PRKG2 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.