Human Recombinant HSP90 alpha (His tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
H36-50H-1000 H36-50H-20 H36-50H-100
77558-168EA 4068 USD
77558-168 77558-170 77558-166
Human Recombinant HSP90 alpha (His tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length human HSP90 alpha was expressed by baculovirus in Sf9 insect cells using a C-terminal His tag.