Human Recombinant JAK2 (JH2) (V617F W659A W777A F794H, GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
J02-16G-100 J02-16G-1000 J02-16G-20
77558-640EA 833 USD
77558-640 77558-642 77558-644
Human Recombinant JAK2 (JH2) (V617F W659A W777A F794H, GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant Human mutant JAK2 JH2 domain (V617F W659A W777A F794H) (536 to 812) was expressed by Baculovirus in Sf9 insect cells using a N to terminal GST tag.