Mouse Recombinant IDO2, (GST Tag) (from Baculovirus (Insect cells))
Supplier: Sino Biological
I212-30G-100 I212-30G-1000 I212-30G-20
77558-546EA 688 USD
77558-546 77558-548 77558-550
Mouse Recombinant IDO2, (GST Tag) (from Baculovirus (Insect cells))
Proteins and Peptides
Recombinant full length mouse IDO2 was expressed by Baculovirus in Sf9 insect cells using an N to terminal GST tag.